Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Ultra-Low Attachment Flasks

Catalog No. 07200876 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
7.5 mL
Culture Area:
25 cm2
Shape:
Rectangular
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Culture Area Shape
07-200-876 7.5 mL 25 cm2 Rectangular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-200-876 Supplier Corning™ Supplier No. 3815

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.

  • 25 to 75 cm2 available growth area
  • Manufactured from optically clear virgin polystyrene
  • Ultra-Low Attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation
  • Nonpyrogenic

Order Info

Sterilized by gamma radiation; certified nonpyrogenic

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 0.25 oz.
Closure Color Orange
Closure Material HDPE
Capacity (Metric) 7.5 mL
Material Polystyrene
For Use With (Application) For pipetting or scraping
Neck Style Canted
No. per Case 24/Cs.
No. per Pack 6/Pk.
Shape Rectangular
Surface Coating Ultra-low Attachment
Graduated Yes
Sterility Sterile
Bottom Shape Rectangular
Closure Type Vented
Culture Area 25 cm2
Show More Show Less

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.