Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Ultra-Low Attachment Flasks
GREENER_CHOICE

Catalog No. 10320171 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
7.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
10-320-171 7.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-320-171 Supplier Corning™ Supplier No. 4616
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.

  • Manufactured from optically clear virgin polystyrene
  • Ultra-Low Attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 0.25 oz.
Closure Color Orange
Closure Material HDPE
Color Clear
Capacity (Metric) 7.5 mL
Material Polystyrene
For Use With (Application) For Pipetting or Scraping
Neck Style Canted
No. per Case 24/Cs.
No. per Pack 6/Pk.
Product Type Cell Culture Flask
Shape Rectangular
Surface Coating Ultra-low Attachment
Porosity 0.2 μm
Disposable Single-use
Graduated Yes
Sterility Sterile
Bottom Shape Rectangular
Closure Type Vented
Culture Area 25 cm²
Surface Treatment Ultra-low Attachment
Volume (Metric) Working 5 to 7.5 mL
Closure Size 20 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.