Learn More
Alkali Scientific Ultra Low Attachment ULA Cell
SDP
Catalog No. 503027221
Encompass_Preferred
Ultra Low Attachment (ULA) cell culture flasks are culture vessels that provides an optimized enviroment for 3D cell culture The culture vessel which is effective for the formation of spheroids of animal or any mamalian cells does not require any special incubation techniques and thus 3D cell culture can easily be implemented in the same way as conventional 2D culture Grow high quality 3D clusters on a wide range of formats from plates dishes and flasks Notably the 96 well plates are optimized commonly used to form a single spheroid Ultra Low Attachment Flasks provides an optimized environment for 3D cell culture Our high quality culture vessels are extremely effective for the culturing of spheroid cells Alkali Scientifics ULA vessesls feature a proprietary covalently bound hydrogel layer that will minimizes cell attachment protein absorption and cellular activation Our Ultra-Low attachment plastics are manufactured from optically clear virgin polystyrene and are supplied with l
Catalog No. 50-302-7221 Supplier Alkali Scientific Supplier No. CFF75
Ultra Low Attachment (ULA) cell culture flasks are culture vessels that provides an optimized enviroment for 3D cell culture The culture vessel which is effective for the formation of spheroids of animal or any mamalian cells does not require any special incubation techniques and thus 3D cell culture can easily be implemented in the same way as conventional 2D culture Grow high quality 3D clusters on a wide range of formats from plates dishes and flasks Notably the 96 well plates are optimized commonly used to form a single spheroid Ultra Low Attachment Flasks provides an optimized environment for 3D cell culture Our high quality culture vessels are extremely effective for the culturing of spheroid cells Alkali Scientifics ULA vessesls feature a proprietary covalently bound hydrogel layer that will minimizes cell attachment protein absorption and cellular activation Our Ultra-Low attachment plastics are manufactured from optically clear virgin polystyrene and are supplied with l