Learn More
PHCbi VIP ECO Large Volume Ultra-Low Freezer
Catalog No. 502720060
Encompass_Preferred
The VIP ECO -86°C ultra-low temperature freezer provides high volume, long term sample storage capability within an optimum footprint. Natural refrigerants minimize energy consumption, reducing environmental impact. Freezer operation is optimally managed by effectively balancing temperature performance and energy management.Effective capacity: 29.8 cu.ft. | 845 LCooling Performance: -86℃
Catalog No. 50-272-0060 Supplier PHCbi Supplier No. MDFDU901VHAPA
Add to Cart
Add to Cart
Request a Quote
The VIP ECO -86°C ultra-low temperature freezer provides high volume, long term sample storage capability within an optimum footprint. Natural refrigerants minimize energy consumption, reducing environmental impact. Freezer operation is optimally managed by effectively balancing temperature performance and energy management.Effective capacity: 29.8 cu.ft. | 845 LCooling Performance: -86℃