Learn More
Hach Company Ultra Low Range CL17sc Kit
Catalog No. 5000207443 Shop All Hach Company Products
Encompass
Ultra Low Range CL17sc Colorimetric Chlorine Analyser with Pressure Regulator Installation KitULR CL17sc Chlorine Analyser-Pressure Regulator Installation Kit-and User Manual User provides screws and/or anchors for mounting
Catalog No. 50-002-07443 Supplier Hach Company Supplier No. 9790300
Add to Cart
Add to Cart
Request a Quote
Ultra Low Range CL17sc Colorimetric Chlorine Analyser with Pressure Regulator Installation KitULR CL17sc Chlorine Analyser-Pressure Regulator Installation Kit-and User Manual User provides screws and/or anchors for mounting