Learn More
PHCbi Ultra-low temperature Freezer
Supplier:  PHCbi MDFDU702VXCPA
Encompass_Preferred
PHCbi brand 25.7 cu.ft (729L) ultra-low temperature freezer features a dual cooling system to offer our highest level of sample protection through the use of two independent refirgeration systems.Temperature Range'-50 °C to -86° C
Catalog No. 50-272-0061
Only null left
Add to Cart
Request a Quote