Learn More
Thermco Products Inc Single Pt100 digital thermometer with probe, -100 deg./300 deg.C & F x 0.1C&F, Certified at -80 deg.C, -20 deg.C, 0 deg.C, 4 deg.C, 25 deg.C, 37 deg.C
Catalog No. NC0518871
Encompass_Preferred
PRECISION Handheld Pt100 Digital Thermometer for the verification and calibration of liquid-in-glass, digital and dial bi-metal thermometer; as well as laboratory instruments such as freezers, refrigerators, incubators, ovens, etc. The PRECISION - Pt100’s are traceable to NIST Standards and is furnished with a certificate proving traceability.
Catalog No. NC0518871 Supplier Thermco Products Inc Supplier No. ACCD370PUFC
PRECISION Handheld Pt100 Digital Thermometer for the verification and calibration of liquid-in-glass, digital and dial bi-metal thermometer; as well as laboratory instruments such as freezers, refrigerators, incubators, ovens, etc. The PRECISION - Pt100’s are traceable to NIST Standards and is furnished with a certificate proving traceability.