Learn More
World Precision Instrument Microinjection Syringe Pump Only-UMP3
SDP
Catalog No. NC1651075
Encompass
With WPI's MICRO2T SMARTouch™ controller the UMP3 offers an intuitive, user-friendly interface . The UMP3 may be mounted to either a stereotaxic frame or a micro manipulator via its built-in mounting bar. An optional foot switch is available.
To learn more about our warranty options, click here
Catalog No. NC1651075 Supplier World Precision Instrument Supplier No. UMP3
With WPI's MICRO2T SMARTouch™ controller the UMP3 offers an intuitive, user-friendly interface . The UMP3 may be mounted to either a stereotaxic frame or a micro manipulator via its built-in mounting bar. An optional foot switch is available.
To learn more about our warranty options, click here