Learn More
Research Products International Corp Ultra Micro Tip, 10ul, Fits Benchmate 0.5-10ul; Pipetman P-2; Pipetman P-10, Autoclavable, Non-Sterile, Bulk, 1000 per Case
SDP
Catalog No. 50212218
Encompass
Our filter pipet tips effectively trap aerosol contaminates when using air displacement pipettors. Ideal for a wide variety of applications including DNA amplification, forensics, tissue culture and protocols that involve pipetting radioactive solutions. The special inert filter is non-self sealing and allows sample recovery in event of over pipetting. Tips are designed for all popular micro-pipettors.
Catalog No. 50-212-218 Supplier Research Products International Corp Supplier No. 141206
Add to Cart
Add to Cart
Our filter pipet tips effectively trap aerosol contaminates when using air displacement pipettors. Ideal for a wide variety of applications including DNA amplification, forensics, tissue culture and protocols that involve pipetting radioactive solutions. The special inert filter is non-self sealing and allows sample recovery in event of over pipetting. Tips are designed for all popular micro-pipettors.