Learn More
Cole-Parmer Thermo Scientific 8302BNUMD ROSS Ultra pH/ATC Probe, DJ/Glass; BNC/MiniDIN
Catalog No. NC1483947 Shop All Cole-Parmer Products
Encompass
This glass body probe builds on the ROSS technology for greater stability, reliability and renowned accuracy, along with temperature response. Its less long-term drift saves time from recalibrating and is double junction with a mini-DIN temperature compensation. It comes with a two-year warranty.
Catalog No. NC1483947 Supplier Cole-Parmer Supplier No. 0571828
This glass body probe builds on the ROSS technology for greater stability, reliability and renowned accuracy, along with temperature response. Its less long-term drift saves time from recalibrating and is double junction with a mini-DIN temperature compensation. It comes with a two-year warranty.