Learn More
World Precision Instrument Ultra Precise Manipulator Arm, 3-Axis, Right handed
SDP
Catalog No. 502539189
Encompass
Ultra-Precise Stereotaxics
  • Ultra-Precise Manipulators are ideal for use in mice and rats where high-accuracy positioning is required.
  • Up to 80mm of travel is available in all directions with up to 90° angle adjustment in either the anterior-posterior or medial-lateral planes.
Catalog No. 50-253-9189 Supplier World Precision Instrument Supplier No. 505223
Ultra-Precise Stereotaxics
  • Ultra-Precise Manipulators are ideal for use in mice and rats where high-accuracy positioning is required.
  • Up to 80mm of travel is available in all directions with up to 90° angle adjustment in either the anterior-posterior or medial-lateral planes.