Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies Ultra Pure Water for CE, 500 mL, Capillary Electrophoresis, Ultra Pure

Catalog No. NC3369964
Encompass

Agilent Technologies Ultra Pure Water for CE is a high-grade reagent designed specifically for demanding Capillary Electrophoresis (CE) applications. This 500 mL volume of ultra-pure water is essential for preparing buffers, samples, and rinsing CE systems, which helps ensure optimal performance and reliable analytical results. The product minimizes contaminants, making it a critical component in analytical chemistry laboratories.

  • Volume: 500 mL
  • Application: Capillary Electrophoresis (CE)
  • Purity level: Ultra pure
  • Identified uses: Reagents and standards for analytical chemistry laboratory
  • Hazard statements: No known significant effects or critical hazards
  • Signal word: No signal word

Catalog No. NC3369964 Supplier Agilent Technologies Supplier No. 50628578
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agilent Technologies Ultra Pure Water for CE is a high-grade reagent designed specifically for demanding Capillary Electrophoresis (CE) applications. This 500 mL volume of ultra-pure water is essential for preparing buffers, samples, and rinsing CE systems, which helps ensure optimal performance and reliable analytical results. The product minimizes contaminants, making it a critical component in analytical chemistry laboratories.

  • Volume: 500 mL
  • Application: Capillary Electrophoresis (CE)
  • Purity level: Ultra pure
  • Identified uses: Reagents and standards for analytical chemistry laboratory
  • Hazard statements: No known significant effects or critical hazards
  • Signal word: No signal word

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.