Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ultra Purified Water, HPLC Grade, Inventus Biotech
SDP

Catalog No. 21520136
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-136 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-136 Supplier Cen-Med Enterprises Supplier No. 353319HPLC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HPLC Grade Water Ultra Purified for analytical and preparative separations. Inventus Biotech HPLC grade reagents contain no impurities to produce spurious peaks in a chromatogram baseline.

  • HPLC Grade Water
  • Ultra Purified
  • Sold in Amber Glass Bottles
  • Case of 4 x 4L

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI 15377
IUPAC Name water
SMILES O

Specifications

Grade HPLC
Packaging Amber Glass Bottle
Quantity 4 x 4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.