Learn More
World Precision Instrument Ultra Quiet Intracellular Amplifier
SDP
Catalog No. 502538507
Encompass
High quality, intracellular amplifier perfect for students
Electro 705, a battery operated, low noise, wide band electrometer preamplifier, is designed for intracellular voltage measurement. Two 705’s can be linked together to form a high impedance differential electrometer pair. Each instrument includes a miniature gold plated active probe to which a microelectrode can be attached using the WPI microelectrode holder supplied.
Catalog No. 50-253-8507 Supplier World Precision Instrument Supplier No. SYS705
High quality, intracellular amplifier perfect for students
Electro 705, a battery operated, low noise, wide band electrometer preamplifier, is designed for intracellular voltage measurement. Two 705’s can be linked together to form a high impedance differential electrometer pair. Each instrument includes a miniature gold plated active probe to which a microelectrode can be attached using the WPI microelectrode holder supplied.