Learn More
Spherotech ULTRA RAINBOW CALIBRATION PART
Catalog No. NC2026776
Encompass_Preferred
Ultra Rainbow Calibration Particles, 6 peaks 107/mL 8.0-12.9 µm 2 mL
Catalog No. NC2026776 Supplier Spherotech Supplier No. URCP1002
Ultra Rainbow Calibration Particles, 6 peaks 107/mL 8.0-12.9 µm 2 mL