Learn More
Spherotech ULTRA RAINBOW MIDRANGE 5ML
Catalog No. NC0930168
Encompass_Preferred
UltraRainbow Fluorescent Particles, Mid-Range, 1E7/mL, 3.5-3.9 um, 5 mL
Catalog No. NC0930168 Supplier Spherotech Supplier No. URFP385A
UltraRainbow Fluorescent Particles, Mid-Range, 1E7/mL, 3.5-3.9 um, 5 mL