Learn More
Crystal Chem Inc Ultra Sensitive Rat Insulin ELISA Kit, 96-well, Uses 5 uL per sample
SDP
Supplier:  Crystal Chem Inc 90060
Encompass
The Ultra Sensitive Rat Insulin ELISA is used to quantify levels of mouse insulin in serum, plasma, cell culture, and fluid using only a 5 µL sample volume.Highlights: Kits use only 5 µL sample. Dynamic range of 0.1- 64 ng/mL. High sensitivity of 50 pg/mL using 5 µL sample. For bulk purchases save money with our 10-kit pack offered at ten invidviual kits.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7918
Edge