Learn More
Grainger Ultra Reflective Letter B PK10
Catalog No. 502751550
Encompass_Preferred
The Brady Letter Label is a 3 inch character height label for labeling and identifying wires, cables, and equipment. It has a white background with bold black lettering for high visibility. The label material is vinyl cloth with a permanent acrylic adhesive for adhesion to a variety of surfaces. It can withstand temperatures from -40°F to 250°F. The label comes in a pack of 250 labels. It meets military standards MIL-STD 202G and MIL-STD 810G for durability. The Brady Letter Label provides a clear and durable labeling solution for industrial applications.
Catalog No. 50-275-1550 Supplier Grainger Supplier No. 14R147
The Brady Letter Label is a 3 inch character height label for labeling and identifying wires, cables, and equipment. It has a white background with bold black lettering for high visibility. The label material is vinyl cloth with a permanent acrylic adhesive for adhesion to a variety of surfaces. It can withstand temperatures from -40°F to 250°F. The label comes in a pack of 250 labels. It meets military standards MIL-STD 202G and MIL-STD 810G for durability. The Brady Letter Label provides a clear and durable labeling solution for industrial applications.