Learn More
Grainger Ultra Reflective Letter H PK10
Catalog No. 502753282
Encompass_Preferred
The Brady Letter Label is a 3 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels are useful for identifying wires, panels, components, equipment, and more. They have high adhesion and are resistant to most solvents and weather conditions. The labels come in a pack of 250 and have a temperature range of -40°F to 176°F. Overall, these vinyl letter labels provide durable and customizable identification.
Catalog No. 50-275-3282 Supplier Grainger Supplier No. 14R153
The Brady Letter Label is a 3 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels are useful for identifying wires, panels, components, equipment, and more. They have high adhesion and are resistant to most solvents and weather conditions. The labels come in a pack of 250 and have a temperature range of -40°F to 176°F. Overall, these vinyl letter labels provide durable and customizable identification.