Learn More
Grainger Ultra Reflective Letter H PK10
Catalog No. 502750486
Encompass_Preferred
The Brady Letter Label is a 2 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels come in a pack of 250 and can be used for general purpose identification and labeling applications. With 2 inch tall characters, these labels are good for visibility at a distance. The vinyl material is durable for indoor and outdoor use.
Catalog No. 50-275-0486 Supplier Grainger Supplier No. 14R117
The Brady Letter Label is a 2 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels come in a pack of 250 and can be used for general purpose identification and labeling applications. With 2 inch tall characters, these labels are good for visibility at a distance. The vinyl material is durable for indoor and outdoor use.