Learn More
Grainger Ultra Reflective Letter J PK10
Catalog No. 502753313
Encompass_Preferred
The Grainger 14R155 is a Westward brand 1/4-inch drive click-type torque wrench. It has a torque range of 10 to 150 inch pounds. The wrench has a reversible ratchet head that allows operation in tight spaces. It features a knurled grip handle and a positive lock with spring-loaded pull down lock ring. The wrench meets ASME B107.14M standards and comes with a certificate of calibration traceable to NIST. It has an accuracy of +/- 4% clockwise and +/- 6% counterclockwise. The wrench measures 12.5 inches long.
Catalog No. 50-275-3313 Supplier Grainger Supplier No. 14R155
The Grainger 14R155 is a Westward brand 1/4-inch drive click-type torque wrench. It has a torque range of 10 to 150 inch pounds. The wrench has a reversible ratchet head that allows operation in tight spaces. It features a knurled grip handle and a positive lock with spring-loaded pull down lock ring. The wrench meets ASME B107.14M standards and comes with a certificate of calibration traceable to NIST. It has an accuracy of +/- 4% clockwise and +/- 6% counterclockwise. The wrench measures 12.5 inches long.