Learn More
Grainger Ultra Reflective Letter P PK10
Catalog No. 502751115
Encompass_Preferred
The Brady 802GU1 is a self-laminating vinyl label with a permanent adhesive backing. It measures 2" x 1" and comes in white. The vinyl material is UV, water, chemical, abrasion, and temperature resistant making it suitable for both indoor and outdoor use. It can adhere to a variety of surfaces including plastic, metal, and more. This versatile label works with the Brady BMP41 handheld label printer allowing for on-demand identification and marking. With its durable construction and compatibility with Brady printers, the 802GU1 provides a labeling solution across many industries and applications.
Catalog No. 50-275-1115 Supplier Grainger Supplier No. 14R125
The Brady 802GU1 is a self-laminating vinyl label with a permanent adhesive backing. It measures 2" x 1" and comes in white. The vinyl material is UV, water, chemical, abrasion, and temperature resistant making it suitable for both indoor and outdoor use. It can adhere to a variety of surfaces including plastic, metal, and more. This versatile label works with the Brady BMP41 handheld label printer allowing for on-demand identification and marking. With its durable construction and compatibility with Brady printers, the 802GU1 provides a labeling solution across many industries and applications.