Learn More
Grainger Ultra Reflective Letter Q PK10
Catalog No. 502753283
Encompass_Preferred
The Brady Number Label is a 2.5" x 1" label with 14 characters per line. It is made of white polyester with a permanent acrylic adhesive and has a temperature range of -40°F to 250°F. The label can be used for wire and cable marking, panel and equipment ID, barcode labeling, and more. It has high adhesion and abrasion resistance. The label comes in a pack of 250.
Catalog No. 50-275-3283 Supplier Grainger Supplier No. 14R162
The Brady Number Label is a 2.5" x 1" label with 14 characters per line. It is made of white polyester with a permanent acrylic adhesive and has a temperature range of -40°F to 250°F. The label can be used for wire and cable marking, panel and equipment ID, barcode labeling, and more. It has high adhesion and abrasion resistance. The label comes in a pack of 250.