Learn More
Grainger Ultra Reflective Letter W PK10
Catalog No. 502749995
Encompass_Preferred
The Brady Letter Label 14R132 is a white vinyl label with a permanent acrylic adhesive designed for indoor use on smooth, flat surfaces like metal, glass, plastic, etc. It measures 0.75" high x 1.25" wide and comes in a pack of 250 labels. The labels are suitable for identifying wires, panels, components, equipment, tools, and more. They feature bold black lettering for easy visibility and have a temperature range of -40°F to 176°F. The acrylic adhesive bonds well to low surface energy plastics and the vinyl material resists UV light, chemicals, abrasion, and moisture. This versatile label works for creating durable and legible identification in industrial environments.
Catalog No. 50-274-9995 Supplier Grainger Supplier No. 14R132
The Brady Letter Label 14R132 is a white vinyl label with a permanent acrylic adhesive designed for indoor use on smooth, flat surfaces like metal, glass, plastic, etc. It measures 0.75" high x 1.25" wide and comes in a pack of 250 labels. The labels are suitable for identifying wires, panels, components, equipment, tools, and more. They feature bold black lettering for easy visibility and have a temperature range of -40°F to 176°F. The acrylic adhesive bonds well to low surface energy plastics and the vinyl material resists UV light, chemicals, abrasion, and moisture. This versatile label works for creating durable and legible identification in industrial environments.