Learn More
Labrepco Inc ULTRA SERIES 26 CU. FT. FRIDGE
SDP
Catalog No. NC2172916
Encompass_Preferred
Ultra Series 26 Cu. Ft. Hinged Glass Door Laboratory RefrigeratorSpecifications:-Cu. Ft.: 26-Door(s): One swing glass door, self-closing (energy efficient,double pane vacuum insulated), right hinged, not reversible-Temperature Range: +1°C to +10°C-Exterior Dimensions: 28 3/8" W x 36 1/2" D x 79 3/4" H-Interior Dimensions: 23 3/4" W x 28" D x 52 1/2" H-Alarm: Audible and visual high and low temperature alarms-Shelves: 4 adjustable shelves (adjustable in ½" increments)-Keyed Door Lock-Electrical Requirements: 115V, 60Hz; 6 amps ; 3/8 HP-UL/C-UL listed-Shipping Weight: 355 lbs.
This item is not returnable. View return policy
Catalog No. NC2172916 Supplier Labrepco Inc Supplier No. LHU26HG
Add to Cart
Add to Cart
Request a Quote
Ultra Series 26 Cu. Ft. Hinged Glass Door Laboratory RefrigeratorSpecifications:-Cu. Ft.: 26-Door(s): One swing glass door, self-closing (energy efficient,double pane vacuum insulated), right hinged, not reversible-Temperature Range: +1°C to +10°C-Exterior Dimensions: 28 3/8" W x 36 1/2" D x 79 3/4" H-Interior Dimensions: 23 3/4" W x 28" D x 52 1/2" H-Alarm: Audible and visual high and low temperature alarms-Shelves: 4 adjustable shelves (adjustable in ½" increments)-Keyed Door Lock-Electrical Requirements: 115V, 60Hz; 6 amps ; 3/8 HP-UL/C-UL listed-Shipping Weight: 355 lbs.