Learn More
Braintree Scientific Ultra Sharp Needle, Sterile, 30ga, 1/2"""", 100/bx
SDP
Catalog No. 501955401
Encompass
Ultra-sharp, tri-beveled, anti-coring, stainless steel needle. Translucent, color-coded luer lock hub. Color-coded cap on rigid pack. Autoclavable in rigid pack for use in O.R/procedure packs. Sterile. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5401 Supplier Braintree Scientific Supplier No. NS30G
Ultra-sharp, tri-beveled, anti-coring, stainless steel needle. Translucent, color-coded luer lock hub. Color-coded cap on rigid pack. Autoclavable in rigid pack for use in O.R/procedure packs. Sterile. **Sold for research purposes only and not for human use**