Learn More
Electron Microscopy Sciences Branson Ultra Sonic Bath Model 3800-MTH, 115 V
SDP
Catalog No. 50930701
Encompass_Preferred
With a unique sweep frequency technology to eliminate standing waves, and a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry standard for quality, reliability, and precision cleaning.
Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level and temperature caused by a light or heavy load. This helps ensure more uniform cleaning and consistent performance.
High/low power control adjusts the acoustic energy in the bath and can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.
Temperature settings up to 69°C/156.2°F can be set in Celsius or Fahrenheit and programmed through the front panel for easy access and monitoring. High-temperature visual alarm/auto shut-off provides added safety. Ultrasonic Cleaner with heat and mechanical timer.
Catalog No. 50-930-701 Supplier Electron Microscopy Sciences Supplier No. 71030MTH
With a unique sweep frequency technology to eliminate standing waves, and a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry standard for quality, reliability, and precision cleaning.
Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level and temperature caused by a light or heavy load. This helps ensure more uniform cleaning and consistent performance.
High/low power control adjusts the acoustic energy in the bath and can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.
Temperature settings up to 69°C/156.2°F can be set in Celsius or Fahrenheit and programmed through the front panel for easy access and monitoring. High-temperature visual alarm/auto shut-off provides added safety. Ultrasonic Cleaner with heat and mechanical timer.