Learn More
Electron Microscopy Sciences Branson Ultra Sonic Bath Model 2800-MT, 115 V, 3/4 gal (2.8L), 9.5"x5.5"x4"
SDP
Catalog No. 501899431
Encompass_Preferred
With a unique sweep frequency technology to eliminate standing waves, & a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry std for quality, reliability, & precision cleaning.Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level & temperature caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.Temperature settings up to 69°C/156.2°F can be set in Celsius or Fahrenheit & programmed through the front panel for easy access & monitoring. High-temperature visual alarm/auto shut-off provides added safety. Ultrasonic Cleaner only w/ mechanical timer.Capacity: 3/4 gal (2.8L)Tank: 9.5 in x 5.5 in x 4 in Footprint: 13.5 in x 12 in Height: 11.5 in
Catalog No. 50-189-9431 Supplier Electron Microscopy Sciences Supplier No. 71025MT
With a unique sweep frequency technology to eliminate standing waves, & a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry std for quality, reliability, & precision cleaning.Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level & temperature caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.Temperature settings up to 69°C/156.2°F can be set in Celsius or Fahrenheit & programmed through the front panel for easy access & monitoring. High-temperature visual alarm/auto shut-off provides added safety. Ultrasonic Cleaner only w/ mechanical timer.Capacity: 3/4 gal (2.8L)Tank: 9.5 in x 5.5 in x 4 in Footprint: 13.5 in x 12 in Height: 11.5 in