Learn More
Electron Microscopy Sciences Branson Ultra Sonic Bath Model 2800-MTH, 115 V, 3/4 gal (2.8L), 9.5"x5.5"x4" tank
SDP
Catalog No. 501899432
Encompass_Preferred
With a unique sweep frequency technology to eliminate standing waves, & a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry standard for quality, reliability, & precision cleaning.Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level & temperature caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage. High-temperature visual alarm/auto shut-off provides added safety. w/ heat & mechanical timer.Capacity: 3/4 gal (2.8L)Tank: 9.5"x5.5"x4"Footprint: 13.5"x12"Height: 11.5"
Catalog No. 50-189-9432 Supplier Electron Microscopy Sciences Supplier No. 71025MTH
With a unique sweep frequency technology to eliminate standing waves, & a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry standard for quality, reliability, & precision cleaning.Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level & temperature caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage. High-temperature visual alarm/auto shut-off provides added safety. w/ heat & mechanical timer.Capacity: 3/4 gal (2.8L)Tank: 9.5"x5.5"x4"Footprint: 13.5"x12"Height: 11.5"