Learn More
MSC Quantum Storage Polypropylene Hopper Stacking Bin, Ivory, 6-9/16 in. Width, 14-3/4 in. Length, 7 in. Height, 12/Pk.
Catalog No. 5000510497
Encompass
This polypropylene hopper stacking bin is designed for efficient organization and storage of components and supplies. Featuring a durable construction, this bin is suitable for a variety of industrial and laboratory environments where organized inventory management is required.
  • Stackable design with a wide ledge and anti-slide lock for stability
  • Compatible with louvered panels or rails for hanging storage
  • Resistant to chemicals, water, rust, and corrosion
  • Autoclavable up to 250 °F
  • FDA approved material construction
  • Integrated front, back, and side grips for easy handling
  • Features a large molded front label area and crystal styrene window
  • Resistant to extreme cold
Catalog No. 50-005-10497 Supplier MSC Supplier No. 73088155
This polypropylene hopper stacking bin is designed for efficient organization and storage of components and supplies. Featuring a durable construction, this bin is suitable for a variety of industrial and laboratory environments where organized inventory management is required.
  • Stackable design with a wide ledge and anti-slide lock for stability
  • Compatible with louvered panels or rails for hanging storage
  • Resistant to chemicals, water, rust, and corrosion
  • Autoclavable up to 250 °F
  • FDA approved material construction
  • Integrated front, back, and side grips for easy handling
  • Features a large molded front label area and crystal styrene window
  • Resistant to extreme cold