Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ Soft Grain Kidskin Leather TIG Welder's Gloves

Catalog No. 191300536A
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-0536A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-130-0536A Supplier Black Stallion™ Supplier No. 35KS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aramid Fiber stitching provides spark and abrasion resistance, while the seamless index finger ensures good hand control.

  • Grain kidskin leather construction is soft and comfortable
  • Aramid Fiber stitching provides spark and abrasion resistance
  • Seamless index finger ensures good hand control
  • Designed for TIG welding applications

Specifications

Color White
Material Grain Kidskin Leather
Size Small
For Use With (Application) TIG Welding
Glove Cuff Color Orange
Glove Cuff Style Gauntlet
Series 35K
Length (English) Cuff 4.5 in.
Length (Metric) Cuff 11.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.