Learn More
Chemglass Life Sciences Ultra Tissue Grinder, 15mL, Conical
SDP
Catalog No. 501532428
Encompass_Preferred
Ultra Tissue Grinder, 15mL, Conical. The pestle tips of theUltra Tissue Grinders have been vitrified using a proprietary silica casting process. The result is a glass-like abrasive surface which provides a more aggressive grinding action to enable efficient processing of difficult and fibrous samples. Ultra Tissue Grinders incorporate the same patented closed system as the standard grinders. They are supplied gamma sterilized in an easy-to-open package with an extra cap for storage and transport plus a patient identification label for traceability.
Catalog No. 50-153-2428 Supplier Chemglass Life Sciences Supplier No. CLS5115
Ultra Tissue Grinder, 15mL, Conical. The pestle tips of theUltra Tissue Grinders have been vitrified using a proprietary silica casting process. The result is a glass-like abrasive surface which provides a more aggressive grinding action to enable efficient processing of difficult and fibrous samples. Ultra Tissue Grinders incorporate the same patented closed system as the standard grinders. They are supplied gamma sterilized in an easy-to-open package with an extra cap for storage and transport plus a patient identification label for traceability.