Learn More
Description
This barrier tape is designed for safety and hazard communication.
- Text: Danger Do Not Enter
- Color: Red and white
- Country of Origin: USA
Specifications
Specifications
| Certifications/Compliance | ISO 3 Class 1, OSHA, RoHS |
| Adhesive Type | Non-adhesive |
| Color | Red |
| Format | Tape |
| Material | Polyethylene |
| Product Type | Control Tape |
| Display Text | Donot Enter |
| For Use With (Application) | Used for hanging heavy Plastic sheeting to create temporary walls, taping cart wheels, pipefitting, ductwork, floor marking, color coding |
| Min. Operating Temperature (Metric) | 93.3°C |
| Quantity | 1 Ea. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
