Learn More
MSC Fas-trak Ultra-Trak SE Application System, 6-in-1, Floor Applicator
Catalog No. 502835799
Encompass
The Ultra-Trak SE 6-in-1 Application System is a versatile, battery-powered solution for applying finishes, sealers, cleaners, and maintainers to various floor types. It features a base unit with a pump that dispenses solution based on wheel revolutions, ensuring precise and efficient coverage. The system also includes a detail applicator for precise edge and corner work.
  • Applies floor finishes or sealers.
  • Cleans lightly soiled hard floors.
  • Applies mop-on restorers.
  • Dispenses solution based on wheel revolutions for controlled application.
  • Includes a detail applicator for edges and corners.
  • Designed for water or solvent-based sealers and finishes.
Catalog No. 50-283-5799 Supplier MSC Supplier No. 69796845
Add to Cart
Add to Cart
Request a Quote
The Ultra-Trak SE 6-in-1 Application System is a versatile, battery-powered solution for applying finishes, sealers, cleaners, and maintainers to various floor types. It features a base unit with a pump that dispenses solution based on wheel revolutions, ensuring precise and efficient coverage. The system also includes a detail applicator for precise edge and corner work.
  • Applies floor finishes or sealers.
  • Cleans lightly soiled hard floors.
  • Applies mop-on restorers.
  • Dispenses solution based on wheel revolutions for controlled application.
  • Includes a detail applicator for edges and corners.
  • Designed for water or solvent-based sealers and finishes.