Learn More
MSC Fas-Trak Industries Head Assembly, Ultra-Trak EZ, for Chemical Application
Catalog No. 502838269
Encompass
This head assembly is designed for use with the Ultra-Trak base unit, a chemical application system for the concrete floor industry. It includes one EZ finish pad and one vinyl head cover, enabling efficient application of various coatings.
  • Applies water or solvent-based concrete coatings at 20,000 sq/ft per hour.
  • Applies densifiers, hardeners, finish coats, and sealers at 25,000 sq/ft per hour.
  • Can reduce product waste by up to 20%.
  • Available in 28 in, 36 in, and 48 in head assembly sizes.
Catalog No. 50-283-8269 Supplier MSC Supplier No. 69797165
This head assembly is designed for use with the Ultra-Trak base unit, a chemical application system for the concrete floor industry. It includes one EZ finish pad and one vinyl head cover, enabling efficient application of various coatings.
  • Applies water or solvent-based concrete coatings at 20,000 sq/ft per hour.
  • Applies densifiers, hardeners, finish coats, and sealers at 25,000 sq/ft per hour.
  • Can reduce product waste by up to 20%.
  • Available in 28 in, 36 in, and 48 in head assembly sizes.