Learn More
Cole-Parmer IKA 3725001 T 25 ULTRA-TURRAX High-Speed Homogenizer 115VAC-unit only
Catalog No. NC0821150 Shop All Cole-Parmer Products
Encompass
Choose from five generators with a sample volume capacity range of 0.3 to 2,000 mL. A digital display shows set speed and can be easily adjusted with its large dial. These systems can be used with tough solids or large volume samples. They are made out of 316 stainless steel.
Catalog No. NC0821150 Supplier Cole-Parmer Supplier No. 0473921
Add to Cart
Add to Cart
Choose from five generators with a sample volume capacity range of 0.3 to 2,000 mL. A digital display shows set speed and can be easily adjusted with its large dial. These systems can be used with tough solids or large volume samples. They are made out of 316 stainless steel.