Learn More
Glo Germ Company UVL 1006 - 21 LED UV Flashlight, 3 AAA batteries included
SDP
Catalog No. NC9528554
Encompass_Preferred
Ultra Violet Lamp, this 21 LED flashlight provides a focused beam of UV light that will fluoresce Glo Germ in broad daylight! Featuring a push button that toggles the light on and off, this lamp is anodized black, and comes with a wrist strap. Requires three AAA batteries (included).
Catalog No. NC9528554 Supplier Glo Germ Company Supplier No. U10
Ultra Violet Lamp, this 21 LED flashlight provides a focused beam of UV light that will fluoresce Glo Germ in broad daylight! Featuring a push button that toggles the light on and off, this lamp is anodized black, and comes with a wrist strap. Requires three AAA batteries (included).