Learn More
Thomson Instrument Company Vent Caps for Optimum Growth 250mL Flask and Ultra Yield 500mL Flask, Sterile
SDP
Catalog No. NC1924321
Encompass_Preferred
Designed for use with Optimum Growth 250 mL Flasks, this sterile vent cap features a 0.2 µm PTFE membrane for enhanced aeration while maintaining sterility. Ideal for cell culture and microbial applications requiring gas exchange, it offers reliable performance and secure flask compatibility.Flask Compatibility: 250mLMembrane: PTFEPore Size: 0.2µmSterility (SAL): 10⁻⁶ Made in the USA
Catalog No. NC1924321 Supplier Thomson Instrument Company Supplier No. 899111
Designed for use with Optimum Growth 250 mL Flasks, this sterile vent cap features a 0.2 µm PTFE membrane for enhanced aeration while maintaining sterility. Ideal for cell culture and microbial applications requiring gas exchange, it offers reliable performance and secure flask compatibility.Flask Compatibility: 250mLMembrane: PTFEPore Size: 0.2µmSterility (SAL): 10⁻⁶ Made in the USA