Learn More
Thomson Instrument Company Ultra Yield Bidirectional Transfer Cap For 1.5L & 2.5L Ultra Yield Flasks
SDP
Catalog No. NC1908535
Encompass_Preferred
Bidirectional Transfer Caps for Ultra Yield 1.5 L and 2.5 L flasks create a closed, sterile path to pump culture in or out—ideal for harvesting or transferring to larger fermenters without opening the vessel.Key features
•Bidirectional flow path for sterile harvests and media additions
•Leak-tight screw interface engineered for Ultra Yield 1.5 L/2.5 L flasks
•Closed, single-use design to minimize contamination risk and cleanup
•Ready to connect with common lab tubing and pumps
•Aseptic handling that reduces open-bench transfers and aerosol exposure
• Made in the USA
Catalog No. NC1908535 Supplier Thomson Instrument Company Supplier No. 899136
Bidirectional Transfer Caps for Ultra Yield 1.5 L and 2.5 L flasks create a closed, sterile path to pump culture in or out—ideal for harvesting or transferring to larger fermenters without opening the vessel.Key features
•Bidirectional flow path for sterile harvests and media additions
•Leak-tight screw interface engineered for Ultra Yield 1.5 L/2.5 L flasks
•Closed, single-use design to minimize contamination risk and cleanup
•Ready to connect with common lab tubing and pumps
•Aseptic handling that reduces open-bench transfers and aerosol exposure
• Made in the USA