Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Silicone Tubing, 0.13 in ID, 65 Shore A

Catalog No. NC3363107
Encompass_Preferred

Sani-Tech Ultra-65 silicone tubing is a biopharmaceutical-grade, platinum-cured tubing engineered for ultra-low extractables and extended pump life. The ULTRA-65-125-2 configuration has a 0.13 in (3.18 mm) inner diameter, 0.06 in (1.59 mm) wall thickness, and 65 Shore A hardness, and is supplied on coils for fluid-transfer applications including sterile fill, drug delivery, and pumping.

  • Platinum-cured silicone material.
  • Hardness: 65 Shore A.
  • Inner diameter: 0.13 in (3.18 mm).
  • Wall thickness: 0.06 in (1.59 mm).
  • Available coil lengths: 25 ft, 50 ft, 100 ft.
  • Engineered for ultra-low extractables and extended pump life.
  • Suitable for sterile fill and transfers, drug delivery, media and buffer processing, and chromatography.
  • Tested to USP <88> Class VI, USP <87>, ISO 10993-5, European Pharmacopoeia 3.1.9.

Catalog No. NC3363107 Supplier Accuflow Systems Supplier No. ULTRA651252
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sani-Tech Ultra-65 silicone tubing is a biopharmaceutical-grade, platinum-cured tubing engineered for ultra-low extractables and extended pump life. The ULTRA-65-125-2 configuration has a 0.13 in (3.18 mm) inner diameter, 0.06 in (1.59 mm) wall thickness, and 65 Shore A hardness, and is supplied on coils for fluid-transfer applications including sterile fill, drug delivery, and pumping.

  • Platinum-cured silicone material.
  • Hardness: 65 Shore A.
  • Inner diameter: 0.13 in (3.18 mm).
  • Wall thickness: 0.06 in (1.59 mm).
  • Available coil lengths: 25 ft, 50 ft, 100 ft.
  • Engineered for ultra-low extractables and extended pump life.
  • Suitable for sterile fill and transfers, drug delivery, media and buffer processing, and chromatography.
  • Tested to USP <88> Class VI, USP <87>, ISO 10993-5, European Pharmacopoeia 3.1.9.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.