Learn More
Alkali Scientific UltraAbsorb™
SDP
Supplier:  Alkali Scientific SF5ML
Encompass_Preferred
UltraAbsorb™ is a black vinyl film that can be applied to either the top or bottom of the plate. Minimizes background fluorescence and cross talk between plate wells. Ideal as a back seal for black plates with clear well bottoms for top reading measurements. Absorbs stray light, reducing the background caused by light scatter. Protects light sensitive samples from degradation. Features a single split liner for easy application.
Catalog No. 50-238-9554