Learn More
MINNEBIO LLC Ultra-Active Nuclease (Serratia marcescens Nuclease, EC 3.1.30.2) Benzonase Alternative Purity 98%10 kU.
Supplier:  MINNEBIO LLC MBPN0110KU
Encompass_Preferred
Ultra-Active Nuclease is extensively utilized across biopharmaceutical and academic research settings for its ability to streamline nucleic acid removal and improve sample processing. Beyond standard cell lysate applications, it enhances protein purification workflows by reducing nucleic acid contamination, facilitates viral vector and vaccine production by maintaining low-viscosity solutions, and supports antibody development by ensuring cleaner preparations for downstream analyses. Its broad compatibility allows use under diverse conditions, including high-salt buffers, detergents, or partially denatured samples, making it highly versatile for complex biological systems. Widely trusted by leading pharmaceutical companies, top research institutes, and university laboratories worldwide, this enzyme delivers reproducible results, contributes to higher assay accuracy, and reduces processing time in critical experimental and manufacturing workflows.
Catalog No. NC2831144