Learn More
MSC ULTRABAKE FLANGELESS PLUG
Catalog No. 5000788469
Encompass
Ultrabake™ flangeless plugs, UFP-SH Series, feature a tapered design and a hollow core, enabling a single plug to fit various threaded and non-threaded holes. They are ideal for high-temperature applications requiring temporary masking.
  • Recommended for powder coating, e-coating, plating & anodizing
  • Withstands temperatures up to 600°F (315°C)
  • Protects threaded holes without masking the surrounding area
  • Reusable
Catalog No. 50-007-88469 Supplier MSC Supplier No. 30544738
Ultrabake™ flangeless plugs, UFP-SH Series, feature a tapered design and a hollow core, enabling a single plug to fit various threaded and non-threaded holes. They are ideal for high-temperature applications requiring temporary masking.
  • Recommended for powder coating, e-coating, plating & anodizing
  • Withstands temperatures up to 600°F (315°C)
  • Protects threaded holes without masking the surrounding area
  • Reusable