Learn More
Electron Microscopy Sciences UltraBed Low Viscosity Kit 2x100 mL
SDP
Catalog No. 50980410
Encompass_Preferred
The kit consists of two components, equal amount, very low viscosity, (~65cps), in convenient dispenser top bottles. To prepare the Embedding Resin, just mix equal amounts of the two components. The resin readily infiltrates into specimen and polymerizes to a clear, hard block overnight at 60°C. Kit comes with 100 mL each component A and component B.
Catalog No. 50-980-410 Supplier Electron Microscopy Sciences Supplier No. 14310
The kit consists of two components, equal amount, very low viscosity, (~65cps), in convenient dispenser top bottles. To prepare the Embedding Resin, just mix equal amounts of the two components. The resin readily infiltrates into specimen and polymerizes to a clear, hard block overnight at 60°C. Kit comes with 100 mL each component A and component B.