Learn More
Beckman Coulter BOTTLES PC 1-1/2 X 4 (BOX-6) 70ML
Catalog No. NC9708359
Encompass_Preferred
Polycarbonate Bottle (No Cap), 70mL, 38x102mm, Quantity: 6/Box
Catalog No. NC9708359 Supplier Beckman Coulter Supplier No. 355655
Polycarbonate Bottle (No Cap), 70mL, 38x102mm, Quantity: 6/Box