Learn More
Thermo Scientific™ Ultrac™ Adapters
Catalog No. 053759 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
12 x 75 mm
Catalog No. Capacity
05-375-9 12 x 75 mm
1 options
Catalog No. 05-375-9 Supplier Thermo Scientific™ Supplier No. 072260F
For use with Centrifuges from Thermo Scientific
Adapters for models Centra™-CL3/CL3R, -GP8/GP8R; HN-SII; EXD and K; and discontinued Centra-MP4/MP4R. Note: rotors require 05-375-60 Shaft Adapter when used on Centra-CL3/-CL3R.
  • With stainless-steel handles for easy loading
  • Autoclavable plastic construction provides full support along length of tube
  • Numbered tube slots for easy identification
  • Withstand centrifugal forces to 50,000×G
TRUSTED_SUSTAINABILITY
Specifications
Capacity 12 x 75 mm