Learn More
Electron Microscopy Sciences Ultraclean Slide
SDP
Catalog No. 5031437
Encompass_Preferred
UltraClean substrate provides users with a pristine glass surface which can be used in any coating process compatible with glass such as silanization and polylysine. Manufactured from the highest quality low fluorescence glass. UltraClean substrates are manufactured in a precise and controlled process incorporating HEPA filtration. This ensures a clean and consistent surface for optimum performance in microarray applications. Flatness is <10 um variation. Printable area: 20 x 70 mm.
Catalog No. 50-314-37 Supplier Electron Microscopy Sciences Supplier No. 63472UC
UltraClean substrate provides users with a pristine glass surface which can be used in any coating process compatible with glass such as silanization and polylysine. Manufactured from the highest quality low fluorescence glass. UltraClean substrates are manufactured in a precise and controlled process incorporating HEPA filtration. This ensures a clean and consistent surface for optimum performance in microarray applications. Flatness is <10 um variation. Printable area: 20 x 70 mm.