Learn More
Research Products International Corp Ultra Clear Cellophane for Drying Gels, 28cm x 50m Roll
SDP
Catalog No. 50550137
Encompass
The membrane used to air dry gels is the most important component. This ultra clear cellophane provides the optimum porosity to ensure uniform drying and to prevent cracking of gels. RPI Cellophane is autoclavable and commonly used for surface growth of plants,1mold spores, and mushrooms.  Unique pore size allows passage of small molecules, nutrients, and antibiotics for screening purposes.
Outperforms conventional plastic film
Laboratory tested for gel drying
Consistent porosity and quality
Catalog No. 50-550-137 Supplier Research Products International Corp Supplier No. 1080
The membrane used to air dry gels is the most important component. This ultra clear cellophane provides the optimum porosity to ensure uniform drying and to prevent cracking of gels. RPI Cellophane is autoclavable and commonly used for surface growth of plants,1mold spores, and mushrooms.  Unique pore size allows passage of small molecules, nutrients, and antibiotics for screening purposes.
Outperforms conventional plastic film
Laboratory tested for gel drying
Consistent porosity and quality