Learn More
GENTECH SOLUTIONS LLC UltraClear Centrifuge Tube, 50mL, Green Caps
Catalog No. NC3728763
Encompass_Preferred
" EliteChem Our centrifuge tubes combine exceptional clarity, outstanding performance, and contaminant-free construction. Designed with precision, they feature in-molded graduation markings that meet ASTM El272 Class A standards, ensuring accuracy in every measurement and further reduce our footprint on the environment to a minimum.Crafted from 100% USP Class VI virgin medical-grade resin, these tubes boast ultra-low teachable metal content for superior safety. Our walls are up to 25% thicker than standard tubes, they withstand 200% higher G-forcers, delivering unmatched durability and reliability. Pair them with our vibrant selection of colored cap options for a seamless fit with most lab equipment and enhanced organization."
Catalog No. NC3728763 Supplier GENTECH SOLUTIONS LLC Supplier No. 1310111011
" EliteChem Our centrifuge tubes combine exceptional clarity, outstanding performance, and contaminant-free construction. Designed with precision, they feature in-molded graduation markings that meet ASTM El272 Class A standards, ensuring accuracy in every measurement and further reduce our footprint on the environment to a minimum.Crafted from 100% USP Class VI virgin medical-grade resin, these tubes boast ultra-low teachable metal content for superior safety. Our walls are up to 25% thicker than standard tubes, they withstand 200% higher G-forcers, delivering unmatched durability and reliability. Pair them with our vibrant selection of colored cap options for a seamless fit with most lab equipment and enhanced organization."