Learn More
GENTECH SOLUTIONS LLC UltraClear Digestion Tube, 50mL, Blue Caps
Catalog No. NC3728761
Encompass_Preferred
EliteChem Our UltraClear™ digestion tubes are crafted to deliver superior clarity and quality, surpassing any other digestion vessel available in the industry. In addition to their outstanding transparency, these tubes feature redesigned in-molded graduation markings that meet ASTM El272-02 Class A standards, ensuring easy readability. Made from proprietary 100% virgin-grade resin with ultra-low teachable metal metal content, these tubes provide exceptional performance. Each case includes a Certificate of Analysis and Volume Accuracy. When paired with our wide range of colored caps, they integrate with laboratoy equipment, offering enhanced organization and seamless functionality.
Catalog No. NC3728761 Supplier GENTECH SOLUTIONS LLC Supplier No. 1300111012
EliteChem Our UltraClear™ digestion tubes are crafted to deliver superior clarity and quality, surpassing any other digestion vessel available in the industry. In addition to their outstanding transparency, these tubes feature redesigned in-molded graduation markings that meet ASTM El272-02 Class A standards, ensuring easy readability. Made from proprietary 100% virgin-grade resin with ultra-low teachable metal metal content, these tubes provide exceptional performance. Each case includes a Certificate of Analysis and Volume Accuracy. When paired with our wide range of colored caps, they integrate with laboratoy equipment, offering enhanced organization and seamless functionality.